7-Amino-1-tetralon

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-alpha-tetralone, 7-Amino-3,4-dihydro-1(2H)-naphthalenone

OR60056-1G OR60056-500MG
APOSOR60056-1GEA 147 EUR
APOSOR60056-1G APOSOR60056-500MG
7-Amino-1-tetralon
7-Amino-1-tetralon
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369817
CAS-Nummer: 22009-40-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR