3-(1,3-Benzodioxol-5-yl)-3-oxopropanenitrile

Lieferant: Apollo Scientific
Achtung

OR60057-1G OR60057-5G
APOSOR60057-1GEA 216 EUR
APOSOR60057-1G APOSOR60057-5G
3-(1,3-Benzodioxol-5-yl)-3-oxopropanenitrile
3-(1,3-Benzodioxol-5-yl)-3-oxopropanenitrile
Formel: C₁₀H₇NO₃
Molecular Weight: 189.17 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08558157
CAS-Nummer: 96220-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR