2-Chlorothiazole-5-carbonitrile

Lieferant: Apollo Scientific
Gefahr

OR60059-1G OR60059-500MG
APOSOR60059-1GEA 361 EUR
APOSOR60059-1G APOSOR60059-500MG
2-Chlorothiazole-5-carbonitrile
2-Chlorothiazole-5-carbonitrile
Formel: C₄HClN₂S
Molecular Weight: 144.5821 g/mol
Schmelzpunkt: 52…57 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03012290
CAS-Nummer: 51640-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR