2-Bromo-3-(bromomethyl)pyridine

Lieferant: Apollo Scientific
Achtung

OR60069-1G OR60069-250MG
APOSOR60069-1GEA 270 EUR
APOSOR60069-1G APOSOR60069-250MG
2-Bromo-3-(bromomethyl)pyridine
2-Bromo-3-(bromomethyl)pyridine
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276299
CAS-Nummer: 94446-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR