Isochinolin-6-amin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Aminoisoquinoline
OR60070-1G
OR60070-5G
OR60070-10G
APOSOR60070-1GEA
21.9
EUR
APOSOR60070-1G
APOSOR60070-5G
APOSOR60070-10G
Isochinolin-6-amin 97%
Isochinolin-6-amin
Formel:
C₉H₈N₂ Molecular Weight: 144,1732 g/mol Schmelzpunkt: 214…216 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114862 CAS-Nummer: 23687-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...