8-Isochinolinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinolin-8-ylboronic acid
OR60072-1G
OR60072-250MG
APOSOR60072-1GEA
263
EUR
APOSOR60072-1G
APOSOR60072-250MG
8-Isochinolinboronsäure
8-Isoquinolineboronic acid
Formel:
C₉H₈BNO₂ Molecular Weight: 172.9800 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11044913 CAS-Nummer: 721401-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...