4,7-Dibrom-2,1,3-benzothiadiazol
Lieferant: Apollo Scientific
|
Gefahr
|
OR60087-5G
OR60087-25G
APOSOR60087-5GEA
29
EUR
APOSOR60087-5G
APOSOR60087-25G
4,7-Dibrom-2,1,3-benzothiadiazol
4,7-Dibromo-2,1,3-benzothiadiazole
Formel:
C₆H₂Br₂N₂S Molecular Weight: 293,97 g/mol Schmelzpunkt: 186…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00658844 CAS-Nummer: 15155-41-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...