7-Brom-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR60094-1G
OR60094-5G
APOSOR60094-1GEA
21.9
EUR
APOSOR60094-1G
APOSOR60094-5G
7-Brom-1H-indazol
7-Bromo-1H-indazole
Formel:
C₇H₅BrN₂ Molecular Weight: 197,0300 g/mol Schmelzpunkt: 126…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371564 CAS-Nummer: 53857-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...