N-Boc-L-valin 97%
Lieferant: Apollo Scientific
APOSOR60097-100GEA
61.1
EUR
APOSOR60097-100G
APOSOR60097-500G
APOSOR60097-1KG
N-Boc-L-valin 97%
N-Boc-L-valin
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 217,3 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065605 CAS-Nummer: 13734-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...