4-Chloro-5-nitropyridin-2(1H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-hydroxy-5-nitropyridine

OR60104-1G OR60104-250MG
APOSOR60104-1GEA 78.6 EUR
APOSOR60104-1G APOSOR60104-250MG
4-Chloro-5-nitropyridin-2(1H)-one
4-Chloro-5-nitropyridin-2(1H)-one
Formel: C₅H₃ClN₂O₃
Molecular Weight: 174,5400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11046303
CAS-Nummer: 850663-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR