1-Methyl-1H-indazol-4-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-Methyl-1H-indazol-4-yl)boronic acid
OR60109-1G
OR60109-250MG
OR60109-5G
APOSOR60109-1GEA
79.9
EUR
APOSOR60109-1G
APOSOR60109-250MG
APOSOR60109-5G
1-Methyl-1H-indazol-4-boronsäure
1-Methyl-1H-indazole-4-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9803 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09870045 CAS-Nummer: 1001907-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...