1H-Indazol-4-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1H-Indazol-4-yl)boronic acid, 1H-indazole-4-boronic acid
OR60110-100MG
OR60110-250MG
APOSOR60110-100MGEA
21.9
EUR
APOSOR60110-100MG
APOSOR60110-250MG
1H-Indazol-4-boronsäure
1H-Indazole-4-boronic acid
Formel:
C₇H₇BN₂O₂ Molecular Weight: 161,9500 g/mol Schmelzpunkt: 166…168 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09878901 CAS-Nummer: 1023595-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...