5-Brom-3-iod-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR60116-1G OR60116-250MG OR60116-5G
APOSOR60116-1GEA 75.7 EUR
APOSOR60116-1G APOSOR60116-250MG APOSOR60116-5G
5-Brom-3-iod-1H-indazol
5-Bromo-3-iodo-1H-indazole
Formel: C₇H₄BrIN₂
Molecular Weight: 322,9300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07781637
CAS-Nummer: 459133-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR