1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR60132-1G OR60132-250MG
APOSOR60132-1GEA 58 EUR
APOSOR60132-1G APOSOR60132-250MG
1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01330895
CAS-Nummer: 92847-41-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR