4-Methylpyridine-2-carboxamide

Lieferant: Apollo Scientific

OR60135-1G OR60135-250MG OR60135-500MG
APOSOR60135-1GEA 393 EUR
APOSOR60135-1G APOSOR60135-250MG APOSOR60135-500MG
4-Methylpyridine-2-carboxamide
4-Methylpyridine-2-carboxamide
Formel: C₇H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08062672
CAS-Nummer: 54089-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR