Isochinolin-4-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Formylisoquinoline, Isoquinoline-4-carboxaldehyde

OR60160-1G OR60160-5G
APOSOR60160-1GEA 56.7 EUR
APOSOR60160-1G APOSOR60160-5G
Isochinolin-4-carbaldehyd
Isochinolin-4-carbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
Schmelzpunkt: 101…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00829440
CAS-Nummer: 22960-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR