2-(3-Methylisoxazol-5-yl)acetic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methylisoxazole-5-acetic Acid, 3-Methyl-5-isoxazoleacetic acid

OR60165-5G
APOSOR60165-5GEA 58 EUR
APOSOR60165-5G
2-(3-Methylisoxazol-5-yl)acetic acid 98%
2-(3-Methylisoxazol-5-yl)acetic acid
Formel: C₆H₇NO₃
Molecular Weight: 141,126 g/mol
Schmelzpunkt: 101…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863539
CAS-Nummer: 19668-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR