5-Nitrobenzoxazol

Lieferant: Apollo Scientific

OR60169-1G OR60169-5G
APOSOR60169-1GEA 21.9 EUR
APOSOR60169-1G APOSOR60169-5G
5-Nitrobenzoxazol
5-Nitrobenzoxazole
Formel: C₇H₄N₂O₃
Molecular Weight: 164,1183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01359855
CAS-Nummer: 70886-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR