2-Methyl-5-nitrobenzoxazol

Lieferant: Apollo Scientific
Achtung

OR60170-1G OR60170-25G OR60170-5G
APOSOR60170-1GEA 21.9 EUR
APOSOR60170-1G APOSOR60170-25G APOSOR60170-5G
2-Methyl-5-nitrobenzoxazol
2-Methyl-5-nitrobenzoxazole
Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466748
CAS-Nummer: 32046-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR