4-Bromo-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR60183-1G OR60183-250MG
APOSOR60183-1GEA 21.9 EUR
APOSOR60183-1G APOSOR60183-250MG
4-Bromo-1H-pyrazole-3-carboxylic acid
4-Bromo-1H-pyrazole-3-carboxylic acid
Formel: C₄H₃BrN₂O₂
Molecular Weight: 190.9828 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01459882
CAS-Nummer: 13745-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR