4-Bromoquinolin-8-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-8-hydroxyquinoline

OR60191-1G OR60191-250MG
APOSOR60191-1GEA 81.2 EUR
APOSOR60191-1G APOSOR60191-250MG
4-Bromoquinolin-8-ol
4-Bromoquinolin-8-ol
Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08688609
CAS-Nummer: 139399-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR