9-Bromo-1-nonene 97%

Lieferant: Apollo Scientific
Achtung

OR60223-25G OR60223-5G OR60223-100G
APOSOR60223-25GEA 120 EUR
APOSOR60223-25G APOSOR60223-5G APOSOR60223-100G
9-Bromo-1-nonene 97%
9-Bromo-1-nonene
Formel: C₉H₁₇Br
Molecular Weight: 205,139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037826
CAS-Nummer: 89359-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR