2-Amino-1,3-propandiol 96%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-1,3-dihydroxypropan, Serinol

OR60269-100G OR60269-500G OR60269-1KG
APOSOR60269-100GEA 26.1 EUR
APOSOR60269-100G APOSOR60269-500G APOSOR60269-1KG
2-Amino-1,3-propandiol 96%
2-Amino-1,3-propanediol
Formel: C₃H₉NO₂
Molecular Weight: 91,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051487
CAS-Nummer: 534-03-2
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR