2-(2-Benzoylphenyl)-2-phenylacetonitril 95%

Lieferant: Apollo Scientific

OR6063-1G OR6063-500MG
APOSOR6063-1GEA 323 EUR
APOSOR6063-1G APOSOR6063-500MG
2-(2-Benzoylphenyl)-2-phenylacetonitril 95%
2-(2-Benzoylphenyl)-2-phenylacetonitril
Formel: C₂₁H₁₅NO
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00231976
CAS-Nummer: 127667-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR