3-(2-Bromethoxy)benzaldehyd

Lieferant: Apollo Scientific

Synonyme: 3-(2-Bromoethoxy)benzenecarboxaldehyde

OR6065-10G OR6065-1G OR6065-5G
APOSOR6065-10GEA 800 EUR
APOSOR6065-10G APOSOR6065-1G APOSOR6065-5G
3-(2-Bromethoxy)benzaldehyd
3-(2-Bromoethoxy)benzaldehyde
Formel: C₉H₉BrO₂
Siedepunkt: 126…130 °C/0,2mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 186191-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR