3-(1-Methyl-1H-pyrazol-3-yl)benzonitril

Lieferant: Apollo Scientific

OR6084-1G OR6084-5G
APOSOR6084-1GEA 102 EUR
APOSOR6084-1G APOSOR6084-5G
3-(1-Methyl-1H-pyrazol-3-yl)benzonitril
3-(1-Methyl-1H-pyrazol-3-yl)benzonitril
Formel: C₁₁H₉N₃
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09065009
CAS-Nummer: 207909-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR