4-(2-Methyl-1-imidazolyl)benzonitrile 97%
Lieferant: Apollo Scientific
OR6085-1G
OR6085-5G
APOSOR6085-1GEA
141
EUR
APOSOR6085-1G
APOSOR6085-5G
4-(2-Methyl-1-imidazolyl)benzonitrile 97%
4-(2-Methyl-1-imidazolyl)benzonitrile
Formel:
C₁₁H₉N₃ Molecular Weight: 183,214 g/mol Schmelzpunkt: 138…141 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
122957-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...