5-Amino-1-chlorisochinolin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR6097-1G
OR6097-500MG
APOSOR6097-1GEA
180
EUR
APOSOR6097-1G
APOSOR6097-500MG
5-Amino-1-chlorisochinolin 98%
5-Amino-1-chloroisoquinoline
Formel:
C₉H₇ClN₂ Molecular Weight: 178,6183 g/mol Schmelzpunkt: 158…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258640 CAS-Nummer: 374554-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...