1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR61002-5G
APOSOR61002-5GEA 132 EUR
APOSOR61002-5G
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester 97%
1,3-Dimethyl-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,0918 g/mol
Schmelzpunkt: 107 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05664574
CAS-Nummer: 847818-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR