3-Amino-4-hydroxyphenyl methyl sulphone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-(methylsulfonyl)phenol 2-Hydroxy-5-(methylsulfonyl)aniline, 3-Amino-4-hydroxyphenyl Methyl sulphone, 2-Hydroxy-5-(methylsulphonyl)aniline, 2-Amino-4-(methylsulphonyl)phenol

OR61009-1G OR61009-5G
APOSOR61009-1GEA 65.4 EUR
APOSOR61009-1G APOSOR61009-5G
3-Amino-4-hydroxyphenyl methyl sulphone 95%
3-Amino-4-hydroxyphenyl methyl sulphone
Formel: C₇H₉NO₃S
Molecular Weight: 187.21 g/mol
Schmelzpunkt: 158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00719450
CAS-Nummer: 98-30-6
EINECS: 202-654-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR