5-Bromo-2-methylbenzenesulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Bromo-2-methylbenzene-1-sulphonyl chloride

OR61027-1G OR61027-250MG OR61027-5G
APOSOR61027-1GEA 59.6 EUR
APOSOR61027-1G APOSOR61027-250MG APOSOR61027-5G
5-Bromo-2-methylbenzenesulphonyl chloride 97%
5-Bromo-2-methylbenzenesulphonyl chloride
Formel: C₇H₆BrClO₂S
Molecular Weight: 269,55 g/mol
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06655596
CAS-Nummer: 69321-56-8
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR