3-Amino-6-brom-1-methyl-1H-indazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-1-methyl-1H-indazol-3-amine

OR61034-1G OR61034-250MG
APOSOR61034-1GEA 114 EUR
APOSOR61034-1G APOSOR61034-250MG
3-Amino-6-brom-1-methyl-1H-indazol
3-Amino-6-bromo-1-methyl-1H-indazole
Formel: C₈H₈BrN₃
Molecular Weight: 226.0700 g/mol
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847108
CAS-Nummer: 1214899-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR