Ethyl-1H-indazol-5-carboxylat 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl Indazole-5-carboxylate, Ethyl 1H-indazole-5-carboxylate, Indazole-5-carboxylic acid ethyl ester
OR61035-1G
OR61035-5G
OR61035-10G
APOSOR61035-1GEA
21.9
EUR
APOSOR61035-1G
APOSOR61035-5G
APOSOR61035-10G
Ethyl-1H-indazol-5-carboxylat 95%
Ethyl-1H-indazol-5-carboxylat
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190,20 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426220 CAS-Nummer: 192944-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...