2,4-Dichlor-7H-pyrrolo[2,3-d]pyrimidin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichloro-7-deazapurine
OR61036-1G
OR61036-100G
OR61036-25G
APOSOR61036-1GEA
21.9
EUR
APOSOR61036-1G
APOSOR61036-100G
APOSOR61036-25G
2,4-Dichlor-7H-pyrrolo[2,3-d]pyrimidin 95%
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₆H₃Cl₂N₃ Molecular Weight: 188.01 g/mol Schmelzpunkt: 250…252 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08059278 CAS-Nummer: 90213-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...