3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine, 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester, 2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester
OR61065-5G
APOSOR61065-5GEA
66.7
EUR
APOSOR61065-5G
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester
Formel:
C₁₂H₁₉BN₂O₃ Molecular Weight: 250,10 g/mol Schmelzpunkt: 131…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923427 CAS-Nummer: 893440-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...