Methyl trans-4-aminocyclohexanecarboxylate hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: trans-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride

OR61067-1G OR61067-5G
APOSOR61067-1GEA 21.9 EUR
APOSOR61067-1G APOSOR61067-5G
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
Methyl trans-4-aminocyclohexanecarboxylate hydrochloride
Formel: C₈H₁₅NO₂
Molecular Weight: 193,67 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08274538
CAS-Nummer: 61367-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR