3-Chlor-2-hydrazinopyridin 98%

Lieferant: Apollo Scientific
Achtung

OR6108-100G OR6108-25G OR6108-5G
APOSOR6108-100GEA 241 EUR
APOSOR6108-100G APOSOR6108-25G APOSOR6108-5G
3-Chlor-2-hydrazinopyridin 98%
3-Chlor-2-hydrazinopyridin
Formel: C₅H₆ClN₃
Molecular Weight: 143.58 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01936479
CAS-Nummer: 22841-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR