2-Amino-5-chlorpyrazin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-amino-5-chloropyrazine
OR61102-1G
OR61102-5G
APOSOR61102-1GEA
43.5
EUR
APOSOR61102-1G
APOSOR61102-5G
2-Amino-5-chlorpyrazin
2-Amino-5-chloropyrazine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03423596 CAS-Nummer: 33332-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...