(3-Chloropyrazin-2-yl)methylamine 95%
Lieferant: Apollo Scientific
Synonyme:
2-(Aminomethyl)-3-chloropyrazine
OR61103-1G
OR61103-250MG
OR61103-5G
APOSOR61103-1GEA
377
EUR
APOSOR61103-1G
APOSOR61103-250MG
APOSOR61103-5G
(3-Chloropyrazin-2-yl)methylamine 95%
(3-Chloropyrazin-2-yl)methylamine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038434 CAS-Nummer: 771581-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...