2-Brompyrazin 98%

Lieferant: Apollo Scientific
Achtung

OR61107-1G OR61107-5G OR61107-25G
APOSOR61107-1GEA 36.4 EUR
APOSOR61107-1G APOSOR61107-5G APOSOR61107-25G
2-Brompyrazin 98%
2-Bromopyrazine
Formel: C₄H₃BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD08275680
CAS-Nummer: 56423-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR