5-Brom-3-methoxy-2-pyrazinamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-3-methoxypyrazin-2-amine
OR61108-1G
OR61108-5G
APOSOR61108-1GEA
21.9
EUR
APOSOR61108-1G
APOSOR61108-5G
5-Brom-3-methoxy-2-pyrazinamin 98%
2-Amino-5-bromo-3-methoxypyrazine
Formel:
C₅H₆BrN₃O Molecular Weight: 204,03 g/mol Schmelzpunkt: 135…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08275726 CAS-Nummer: 5900-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...