3-Chlor-5-nitrobenzonitril 98%

Lieferant: Apollo Scientific

OR6111-1G OR6111-5G
APOSOR6111-1GEA 158 EUR
APOSOR6111-1G APOSOR6111-5G
3-Chlor-5-nitrobenzonitril 98%
3-Chlor-5-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
Schmelzpunkt: 84…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00463805
CAS-Nummer: 34662-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR