4,6-Dichloro-5-pyrimidinecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR61112-10G
OR61112-1G
OR61112-5G
APOSOR61112-10GEA
372
EUR
APOSOR61112-10G
APOSOR61112-1G
APOSOR61112-5G
4,6-Dichloro-5-pyrimidinecarboxylic acid
4,6-Dichloro-5-pyrimidinecarboxylic acid
Formel:
C₅H₂Cl₂N₂O₂ Molecular Weight: 192.99 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909611 CAS-Nummer: 87600-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...