Methyl-3-chlor-5-nitrobenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chlor-5-nitrobenzoesäuremethylester
OR6112-1G
OR6112-5G
OR6112-250MG
APOSOR6112-1GEA
125
EUR
APOSOR6112-1G
APOSOR6112-5G
APOSOR6112-250MG
Methyl-3-chlor-5-nitrobenzoat 98%
Methyl-3-chlor-5-nitrobenzoat
Formel:
C₈H₆ClNO₄ Molecular Weight: 215.59 g/mol Siedepunkt: 314 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01529623 CAS-Nummer: 36138-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...