3-Amino-3-(3-bromophenyl)oxetane hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR61147-1G OR61147-5G
APOSOR61147-1GEA 928 EUR
APOSOR61147-1G APOSOR61147-5G
3-Amino-3-(3-bromophenyl)oxetane hydrochloride 97%
3-Amino-3-(3-bromophenyl)oxetane hydrochloride
Formel: C₉H₁₁BrClNO
Molecular Weight: 264.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19381731
CAS-Nummer: 1332920-63-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR