(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,4R)-1-(tert-Butoxycarbonyl)-4-methylpyrrolidine-2-carboxylic acid, (2S,4R)-4-Methylpyrrolidine-2-carboxylic acid, N-BOC protected

OR61167-250MG OR61167-1G
APOSOR61167-250MGEA 208 EUR
APOSOR61167-250MG APOSOR61167-1G
(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid 97%
(2S,4R)-1-[(tert-Butoxy)carbonyl]-4-methylpyrrolidine-2-carboxylic acid
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185988
CAS-Nummer: 364750-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR