5-Bromopyridine-3,4-diamine

Lieferant: Apollo Scientific
Achtung

OR61210-5G OR61210-10G
APOSOR61210-5GEA 224 EUR
APOSOR61210-5G APOSOR61210-10G
5-Bromopyridine-3,4-diamine
5-Bromopyridine-3,4-diamine
Formel: C₅H₆BrN₃
Molecular Weight: 188,03 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01692479
CAS-Nummer: 4635-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR