2-Acetyl-5-bromo-2H-indazole

Lieferant: Apollo Scientific
Achtung

OR61229-1G OR61229-250MG
APOSOR61229-1GEA 183 EUR
APOSOR61229-1G APOSOR61229-250MG
2-Acetyl-5-bromo-2H-indazole
2-Acetyl-5-bromo-2H-indazole
Formel: C₉H₇BrN₂O
Molecular Weight: 239.08 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11100122
CAS-Nummer: 1195623-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR