2-Butyl-5-nitrobenzoxazol

Lieferant: Apollo Scientific

APOSOR61231-5GEA 257 EUR
APOSOR61231-5G APOSOR61231-1G APOSOR61231-10G
2-Butyl-5-nitrobenzoxazol
2-Butyl-5-nitrobenzoxazole
Formel: C₁₁H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05663810
CAS-Nummer: 886360-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR