2-Oxetanemethanamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Aminomethyl)oxetane, Oxetan-2-ylmethanamine

OR61268-1G OR61268-250MG
APOSOR61268-1GEA 493 EUR
APOSOR61268-1G APOSOR61268-250MG
2-Oxetanemethanamine
2-Oxetanemethanamine
Formel: C₄H₉NO
Molecular Weight: 87.12 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD17214171
CAS-Nummer: 882402-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR